missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium hydrogen carbonate, 99.5%, for biochemistry

Product Code. 12645137
500 g
Click to view available options
Quantity:
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
SMILES [K+].OC([O-])=O

Specifications

Melting Point 292.0°C
Quantity 500 g
Linear Formula KHCO3
Merck Index 15773
Solubility Information Clear colorless
IUPAC Name potassium hydrogen carbonate
Formula Weight 100.12
Percent Purity 99.5%
Grade Biochemical
Packaging Plastic Bottle
Chemical Name or Material Potassium hydrogen carbonate
Show More Show Less
Product Identifier
  • Potassium hydrogen carbonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Potassium hydrogen carbonate, 99.5%, for biochemistry >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.