missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydrogen Carbonate, Certified AR for Analysis, Fisher Chemical™
155.00 € - 192.00 €
Chemical Identifiers
CAS | 298-14-6 |
---|---|
Molecular Formula | CHKO3 |
Molecular Weight (g/mol) | 100.11 |
MDL Number | MFCD00011402 |
InChI Key | TYJJADVDDVDEDZ-UHFFFAOYSA-M |
Synonym | potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 |
PubChem CID | 516893 |
ChEBI | CHEBI:81862 |
SMILES | [K+].OC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10568820
|
Fisher Chemical
P/5120/53 |
500 g |
192.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10204210
|
Fisher Chemical
P/5120/60 |
1 kg |
155.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
298-14-6 | |
100.11 | |
TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
516893 | |
[K+].OC([O-])=O |
CHKO3 | |
MFCD00011402 | |
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 | |
CHEBI:81862 |
Specifications
292°C | |
Yellow | |
Solid | |
MFCD00011402 | |
TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
potassium hydrogen carbonate | |
516893 | |
100.12g/mol |
298-14-6 | |
8.2 | |
CHKO3 | |
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 | |
[K+].OC([O-])=O | |
100.11 | |
CHEBI:81862 | |
Plastic powder jar |
Safety and Handling
Product Identifier
- Potassium hydrogen carbonate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title