missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.1M, BAKER ANALYZED, J.T. Baker™

Product Code. 10279480
Click to view available options
Quantity:
1 L
Packaging:
HDPE
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality grade volumetric solution is ready to use for laboratory and specialized industrial applications.

  • Standardized potentiometrically against an NIST Standard Reference Material, Potassium Hydrogen Phthalate
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Boiling Point 100°C
CAS 1310-58-3
Melting Point 0°C
Molecular Formula HKO
Concentration 0.0995–0.1005mol/L
MDL Number MFCD00003553
Density 1.0g/cm3 at 20°C
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Fully miscible in water
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Grade BAKER ANALYZED™
Normality 0.1N
Packaging HDPE
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'