Learn More
Potassium hydroxide, 0.5N Standardized Solution, Thermo Scientific Chemicals
Description
Potassium hydroxide, 0.5N Standardized Solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 L |
Solubility Information | Freely soluble in water. Soluble in ethanol. |
IUPAC Name | potassium;hydroxide |
Formula Weight | 56.11 |
Appearance | Colorless liquid |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Chemical Name or Material | Potassium hydroxide |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
62400667 | Certificate of Analysis | 12/02/2025 | 035595.K2,035595.M4,035595.K7 |
62500025 | Certificate of Analysis | 03/02/2025 | 035595.K2,035595.M4,035595.K7 |
M11L515 | Certificate of Analysis | 17/01/2025 | 035595.K2,035595.M4,035595.K7 |
M11L516 | Certificate of Analysis | 17/01/2025 | 035595.K2,035595.M4,035595.K7 |
Z14K517 | Certificate of Analysis | 26/12/2024 | 035595.K2,035595.M4,035595.K7 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.