Learn More
Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
32.76 € - 261.30 €
Specifications
Chemical Name or Material | Potassium hydroxide |
---|---|
Color | Colorless |
Name Note | 1N solution in water |
Boiling Point | 100.0°C |
CAS | 1310-58-3, 7732-18-5 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10664545
|
Thermo Scientific Acros
278400010 |
1 L | Plastic bottle |
32.76 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10774935
|
Thermo Scientific Acros
278400025 |
2.5 L | Plastic bottle |
46.23 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15109263
|
Thermo Scientific Acros
278400250 |
25 L | Plastic Can |
261.30 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium hydroxide | |
1N solution in water | |
1310-58-3, 7732-18-5 | |
KOH | |
MFCD00003553 | |
01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
56.11 | |
CHEBI:32035 | |
1.05 |
Colorless | |
100.0°C | |
HKO | |
1N, exact strength on the certificate of analysis | |
1814 | |
1.0500g/mL | |
Solubility in water: soluble. | |
[OH-].[K+] | |
14797 | |
56.11 |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Potassium hydroxide, 1N solution in water
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.