missing translation for 'onlineSavingsMsg'
Learn More
Potassium hydroxide, 30% w/v aq. soln., Thermo Scientific Chemicals Product Code.: 11351568

Potassium hydroxide, 30% w/v aq. soln., Thermo Scientific Chemicals

Product Code. 11351568
1 L
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy

Product Code. 11351568

Brand: Thermo Scientific Alfa Aesar 010979.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Color Colorless
Density 1.28
Solubility Information Freely soluble in water. Soluble in ethanol.
Quantity 1 L
IUPAC Name potassium;hydroxide
Formula Weight 56.11
Concentration or Composition (by Analyte or Components) Potassium hydroxide anhydrous: 30.0%; Water: 70.0%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Potassium hydroxide, 30% w/v aqueous solution

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM16L532 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number010979.K2,010979.AP
Lot NumberW27K511 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number010979.K2,010979.AP
Lot NumberW26K509 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number010979.K2,010979.AP
Lot NumberR23K505 Certificate TypeCertificate of Analysis Date04/07/2024 Catalog Number010979.K2,010979.AP
Lot NumberN15K527 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number010979.K2,010979.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium hydroxide, 30% w/v aq. soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.