missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide, ACS Reagent, pellets, ≥85%, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
6 x 500 g
12 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Linear Formula | KOH |
UN Number | 1813 |
Quantity | 2.5 kg |
IUPAC Name | potassium;hydroxide |
Formula Weight | 56.11g/mol |
Percent Purity | ≥85% |
Grade | ACS Reagent |
Vapor Pressure | 1mmHg (719°C) |
Chemical Name or Material | Potassium Hydroxide |
Safety and Handling
Product Identifier
- Potassium hydroxide
- Danger
- Acute toxicity Category 4
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : potassium hydroxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction