missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Alchoholic, Ph. Eur. Grade, 0.5M, Reagecon™
BIOPHARMA

Product Code. 11404809
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Concentration 0.5M
Quantity 1 L
compliance-icons compliance-icons
Product Identifier
  • 0.5M POTASSIUM HYDROXIDE ALCOHOLIC
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ethanol, potassium hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'