missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, ca. 85%, ACS reagent, Thermo Scientific Chemicals

25.02 € - 655.00 €

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10506992
View Documents Promotion Details
Thermo Scientific Acros
424140250
25 g
25.02 €
25g
Please to purchase this item. Need a web account? Register with us today!
10474321
View Documents Promotion Details
Thermo Scientific Acros
424145000
500 g
29.85 €
500g
Please to purchase this item. Need a web account? Register with us today!
10677581
View Documents Promotion Details
Thermo Scientific Acros
424140025
2.5 kg
109.55 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
10073923
View Documents Promotion Details
Thermo Scientific Acros
424140050
5 kg
228.14 €
5kg
Please to purchase this item. Need a web account? Register with us today!
13428578
View Documents Promotion Details
Thermo Scientific Acros
424140251
25 kg
655.00 €
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]

Specifications

White
1310-58-3
ca. 85%
KOH
01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
56.11
ACS Reagent
Pellet
1320.0°C
360.0°C
HKO
MFCD00003553
15, 7761
Solubility in water: 1120g/L (20°C). Other solubilities: 1780g/L water (100°C), soluble in ethanol (400g/L), soluble in ammonia, slightly soluble in ether
[OH-].[K+]
56.11
CHEBI:32035
∼85%
Plastic Bottle
Potassium hydroxide, 85%
Videos
Safety and Handling

Safety and Handling

Danger

missing translation for 'einecsNumber' : 215-181-3

missing translation for 'rtecsNumber' : TT2100000

missing translation for 'tsca' : TSCA

Product Identifier
  • Potassium hydroxide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only