missing translation for 'onlineSavingsMsg'
Learn More
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals Product Code.: 10175723

Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals

Product Code. 10175723
10 kg, Plastic drum
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10175723

Brand: Thermo Scientific Acros 134060100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Boiling Point 1320.0°C
Melting Point 360.0°C
Assay Percent Range ca. 85%
Linear Formula KOH
Fieser 01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258
Merck Index 15, 7761
Solubility Information Solubility in water: 1120g/L (20°C). Other solubilities: 1780g/L water (100°C), soluble in ethanol (400g/L), soluble in ammonia, slightly soluble in ether
Quantity 10 kg
IUPAC Name potassium hydroxide
Formula Weight 56.11
Percent Purity ∼85%
Grade Analytical
Packaging Plastic drum
Physical Form Pellet
Chemical Name or Material Potassium hydroxide, ca. 85 %
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453396 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number134060250,134060010,134060100,134062500,134060025
Lot NumberA0467649 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number134060250,134060010,134060100,134062500,134060025
Lot NumberA0445519 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number134060250,134060010,134060100,134062500,134060025
Lot NumberA0453396 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number134060250,134060010,134060100,134062500,134060025
Lot NumberA0443808 Certificate TypeCertificate of Analysis Date06/07/2022 Catalog Number134060250,134060010,134060100,134062500,134060025
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.