missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, Pure, Solution 48-50%, Liquor, Fisher Chemical
Click to view available options
Quantity:
2.5 L
Specifications
Specifications
pH | 12 |
Color | Colorless |
CAS | 1310-58-3 |
Melting Point | -28.8°C |
Molecular Formula | HKO |
MDL Number | MFCD00003553 |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Quantity | 2.5 L |
SMILES | [OH-].[K+] |
Show More |
Safety and Handling
Product Identifier
- Potassium hydroxide, 30-50% solution in water
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Potassium hydroxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction