missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide solution, Volumetric, 0.5 M KOH in ethanol (0.5N), Honeywell Fluka™
Volumetric, 0.5 M KOH in ethanol (0.5N)
51.18 € - 600.49 €
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
SMILES | [OH-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15694810
|
Honeywell-Fluka
35115-500ML |
500 mL |
51.18 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15664810
|
Honeywell-Fluka
35115-1L |
1 L |
86.48 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15674810
|
Honeywell-Fluka
35115-2.5L |
2.5 L |
173.35 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15614820
|
Honeywell-Fluka
35115-6X500ML |
6 x 500 mL | N/A | ||||||
15604820
|
Honeywell-Fluka
35115-6X1L |
6 x 1 L |
456.13 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15684810
|
Honeywell-Fluka
35115-4X2.5L |
4 x 2.5 L |
600.49 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
CHEBI:32035 |
HKO | |
MFCD00003553 | |
14797 | |
[OH-].[K+] |
Specifications
1310-58-3 | |
KOH | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
[OH-].[K+] | |
56.11 | |
14797 | |
56.11g/mol |
HKO | |
0.5M in ethanol | |
UN2924 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium;hydroxide | |
11°C (51.8°F) | |
CHEBI:32035 | |
Potassium hydroxide solution |
Safety and Handling
P210-P260-P280-P305 + P351 + P338 + P310-P370 + P378-P403 + P235
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium hydroxide solution
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : potassium hydroxide;caustic potash, Toluene, ethanol; ethylalcohol, Hydrazinehydrate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title