missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide solution, 0.5M, Honeywell Fluka™
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
CAS | 1310-58-3 |
Molecular Formula | HKO |
Linear Formula | KOH |
Concentration | 0.5M |
MDL Number | MFCD00003553 |
UN Number | UN1814 |
Density | 1.023g/mL at 20°C |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Quantity | 1 L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'