missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide solution, Volumetric, 0.1 M KOH in ethanol (0.1N), Honeywell Fluka™
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
Linear Formula | KOH |
Concentration | 0.1M in ethanol |
UN Number | UN1170 |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
Quantity | 1 L |
Flash Point | 14°C (57.2°F) |
Formula Weight | 56.11g/mol |
Chemical Name or Material | Potassium hydroxide solution |
Safety and Handling
Product Identifier
- Potassium hydroxide solution
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : ethanol; ethyl alcohol, Toluene, potassium hydroxide; caustic potash, hydrazine
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'