missing translation for 'onlineSavingsMsg'
Learn More
Potassium hydroxide solution, Volumetric, 1 M KOH in ethanol (1N), Honeywell Fluka™ Product Code.: 15634810
compliance-icons compliance-icons

Potassium hydroxide solution, Volumetric, 1 M KOH in ethanol (1N), Honeywell Fluka™

Product Code. 15634810 Shop All Honeywell Fluka Products
1 L
Click to view available options
Quantity:
1 L
6 x 1 L
This item is not returnable. View return policy

Product Code. 15634810

Brand: Honeywell-Fluka 351121L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, 1 M KOH in ethanol (1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693Show MoreShow Less
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
Concentration 1M in ethanol
UN Number UN2924
Quantity 1 L
Flash Point 11°C (51.8°F)
Formula Weight 56.11g/mol
Chemical Name or Material Potassium hydroxide solution
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide,Toluene, Hydrazinehydrate, ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium hydroxide solution, Volumetric, 1 M KOH in ethanol (1N), Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.