missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, Volumetric, 8.0M KOH (8.0N), Honeywell™ Fluka™

Product Code. 15648030
Click to view available options
Quantity:
50 mL
Packaging:
Poly bottle

Volumetric, 8.0 M KOH (8.0N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

pH 14 (at 20°C)
Color Colorless
Boiling Point ∼112°C (1013hPa)
CAS 1310-58-3
Melting Point ∼-6°C
Molecular Formula HKO
Linear Formula KOH
MDL Number MFCD00003553
UN Number UN1814
Density ∽1.510g/cu.cm at 20°C
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Completely soluble in water
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 50 mL
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Odor Odorless
Packaging Poly bottle
Physical Form Liquid
Chemical Name or Material Potassium hydroxide solution, Volumetric, 8.0M KOH (8.0N)
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'