Learn More
Potassium Iodate, Reagent ACS, Powder, +99%, Thermo Scientific Chemicals
51.15 € - 636.17 €
Chemical Identifiers
CAS | 7758-05-6 |
---|---|
Molecular Formula | IKO3 |
Molecular Weight (g/mol) | 214.00 |
MDL Number | MFCD00011406 |
InChI Key | JLKDVMWYMMLWTI-UHFFFAOYSA-M |
Synonym | potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat |
PubChem CID | 23665710 |
IUPAC Name | potassium;iodate |
SMILES | [O-]I(=O)=O.[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10762212
|
Thermo Scientific Acros
418241000 |
100 g | Plastic bottle |
51.15 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10697201
|
Thermo Scientific Acros
418245000 |
500 g | Plastic bottle |
178.89 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10732204
|
Thermo Scientific Acros
418240025 |
2.5 kg | Plastic bottle |
636.17 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7758-05-6 | |
214.00 | |
JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
23665710 | |
[O-]I(=O)=O.[K+] |
IKO3 | |
MFCD00011406 | |
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
potassium;iodate |
Specifications
7758-05-6 | |
5.0 to 8.0 (5% soln. at 25°C) | |
99.4+% | |
KIO3 | |
15, 7763 | |
JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
potassium;iodate | |
23665710 | |
99.4 to 100.4% (Iodometry) | |
560°C | |
Potassium iodate |
White | |
Crystalline Powder and/or Crystals | |
IKO3 | |
MFCD00011406 | |
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
[O-]I(=O)=O.[K+] | |
214.00 | |
214 | |
ACS Reagent | |
3.9300g/mL |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-831-9
missing translation for 'rtecsNumber' : NN1350000
missing translation for 'tsca' : TSCA
- Potassium iodate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only