missing translation for 'onlineSavingsMsg'
Learn More
Potassium iodate solution, 0.05M, Honeywell™ Product Code.: 15684140

Potassium iodate solution, 0.05M, Honeywell™

Product Code. 15684140 Shop All Honeywell Fluka Products
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 15684140

Brand: Honeywell-Fluka 342741L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Volumetric, 0.05 M KIO3 (0.3N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7758-05-6
Quantity 1 L
Molecular Formula IKO3
Linear Formula KIO3
MDL Number MFCD00011406
UN Number NONH for all modes of transport
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES [O-]I(=O)=O.[K+]
IUPAC Name potassium;iodate
Molecular Weight (g/mol) 214.00
PubChem CID 23665710
Formula Weight 214g/mol
Concentration or Composition (by Analyte or Components) 0.05M
Chemical Name or Material Potassium iodate solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium iodate solution, 0.05M, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.