missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodide, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Potassium iodide is a simple iodide salt utilized as a fluorescence quenching agent which acts by collisional quenching of fluorescent substances by the iodide ion. It is also used in the isolation of single stranded DNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 1330°C |
Melting Point | 681°C |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Soluble in water (145g/100ml at 25°C),alcohol (45.5mg/ml),methanol (125mg/ml),acetone (13.1mg/ml at 25°C),and ethanol (19.6mg/ml). |
Formula Weight | 166.01 |
Odor | Odorless |
Refractive Index | 1.677 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'