missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, 99+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10647381
Click to view available options
Quantity:
500 g
1 kg
5 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330.0°C
Melting Point 680.0°C
Color White
pH 6.0 to 9.2 (5% soln. at 25°C)
Physical Form Crystals or Granules
Quantity 500 g
Assay Percent Range 99+%
Linear Formula KI
Fieser 06,227; 11,440
Merck Index 15, 7764
Solubility Information Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Molecular Weight (g/mol) 166.003
Formula Weight 166
Percent Purity ≥99%
Grade ACS Reagent
Loss on Drying 0.2% max. (150°C, 6 hrs)
Vapor Pressure 1mmHg at 745°C
Density 3.13
Chemical Name or Material Potassium iodide, granular
Show More Show Less
compliance-icons
Product Identifier
  • Potassium iodide
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'