missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals

45.12 € - 1816.04 €

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10267523
SDS View Documents
Thermo Scientific Acros
196731000
100 g Glass bottle
45.12 €
100g
Please to purchase this item. Need a web account? Register with us today!
10082610
SDS View Documents
Thermo Scientific Acros
196735000
500 g Glass bottle
154.77 €
500g
Please to purchase this item. Need a web account? Register with us today!
10551451
SDS View Documents
Thermo Scientific Acros
196730025
2.5 kg Plastic bottle
464.31 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
11964211
SDS View Documents
Thermo Scientific Acros
196730100
10 kg Plastic drum
1816.04 €
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7681-11-0
166
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
CHEBI:8346
[K+].[I-]
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide

Specifications

1330.0°C
7681-11-0
Briquettes
IK
06,227; 11,44
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
166
Extra Pure
Potassium iodide
680.0°C
White
98.5% min. (on dried substance) (Argentometry)
KI
15,7764
Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
[K+].[I-]
166
CHEBI:8346
99%
1% max.
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-659-4

Product Identifier
  • Potassium iodide

Signal Word
  • Danger

Hazard Category
  • Specific target organ toxicity Category 1

Hazard Statement
  • H372-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.