missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, ACS, 99.0% min, Thermo Scientific Chemicals

For Use in pHotographic Emulsions, in Analytical Chemistry, As a Feed Additive

53.27 € - 437.18 €

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357347
View Documents
Thermo Scientific Alfa Aesar
011601.22
100 g
53.27 €
100g
Please to purchase this item. Need a web account? Register with us today!
11347347
View Documents
Thermo Scientific Alfa Aesar
011601.36
500 g
169.85 €
500g
Please to purchase this item. Need a web account? Register with us today!
11367347
View Documents
Thermo Scientific Alfa Aesar
011601.A3
2 kg
437.18 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is both a source of iodide and a mild reducing agent. It is used as a component in the electrolyte of dye sensitized solar cells. In the field of synthetic organic chemistry, it is used in the preparation of alkyl or aryl iodo compounds. It is employed as a nucleophilic catalyst in the alkylations with alkyl bromide, chloride, tosylate and mesylate. In the presence of acid it forms hydroiodic acid, which is a strong reducing agent. It is useful in producing Lugol’s solution (or Lugol’s iodne) which finds many applications including starch testing, gram stains, and etching of gold surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7681-11-0
166.003
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide
IK
MFCD00011405
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
CHEBI:8346
[K+].[I-]

Specifications

1330°C
7681-11-0
Granules
IK
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
166.01
Odorless
Hygroscopic
1.33 hPa (1mm Hg)
Potassium iodide, For analysis ACS
681°C
White
99.0% min
MFCD00011405
Soluble in water,alcohol,glycerol,and acetone. Slightly soluble in ether,and ammonia.
[K+].[I-]
166.003
CHEBI:8346
ACS
1.677
White crystalline powder
3.12
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P314-P501c

H372

missing translation for 'einecsNumber' : 213-659-4

missing translation for 'rtecsNumber' : TT2975000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Potassium iodide

Signal Word
  • Danger

Hazard Category
  • Specific target organ toxicity Category 1

Hazard Statement
  • H372-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Potassium iodide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.