missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, Certified AR for Analysis, Fisher Chemical
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Specifications
Specifications
Boiling Point | 1330°C |
Melting Point | 680°C |
CAS | 7681-11-0 |
Color | White |
pH | 6 to 8 |
Physical Form | Solid |
Quantity | 1 kg |
Molecular Formula | IK |
MDL Number | 11405 |
Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide |
Show More |
Safety and Handling
Product Identifier
- Potassium iodide
- Danger
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction