missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
Description
Potassium Iodide, Extra Pure, SLR
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 1330°C |
Melting Point | 680°C |
Color | White |
pH | 6 to 8 |
Physical Form | Solid |
Quantity | 100 g |
Formula Weight | 166g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 1mmHg 745 |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Potassium iodide
- Danger
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'