missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, Puriss., meets analytical spec. of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™
BIOPHARMA

Puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)

83.66 € - 6233.70 €

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]
View More Specs

Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15610670
BIOPHARMA
SDS View Documents
Honeywell-Fluka
03124-250G
250 g
83.66 €
250g
Please to purchase this item. Need a web account? Register with us today!
15600670
BIOPHARMA
SDS View Documents
Honeywell-Fluka
03124-1KG
1 kg
317.75 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15660670
BIOPHARMA
SDS View Documents
Honeywell-Fluka
03124-6X250G
6 x 250 g
384.70 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15640670
BIOPHARMA
SDS View Documents
Honeywell-Fluka
03124-5KG
5 kg
1354.00 €
5kg
Please to purchase this item. Need a web account? Register with us today!
15650670
BIOPHARMA
SDS View Documents
Honeywell-Fluka
03124-6X1KG
6 x 1 kg
1462.38 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15620670
View Documents
Honeywell-Fluka
03124-25KG
25 kg N/A N/A N/A
15630670
View Documents
Honeywell-Fluka
03124-50KG
50 kg N/A N/A N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P305 + P351 + P338

H302-H315-H319

missing translation for 'einecsNumber' : 231-659-4

missing translation for 'rtecsNumber' : TT2975000

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium iodide, Puriss., meets analytical spec. of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.