missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodide, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
6 x 100 g
6 x 250 g
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Boiling Point | 1,323°C |
Melting Point | 681°C (literature) |
CAS Max % | 100.0000% |
Quantity | 6 x 250 g |
Linear Formula | KI |
Formula Weight | 166 |
Percent Purity | >99.5% |
Grade | puriss. p.a. |
Packaging | Poly Bottle |
Density | 3.12 g/cm3 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'