Learn More
Potassium manganate, Thermo Scientific Chemicals
88.04 € - 305.52 €
Chemical Identifiers
CAS | 10294-64-1 |
---|---|
Molecular Formula | K2MnO4 |
Molecular Weight (g/mol) | 197.13 |
MDL Number | MFCD00043089 |
InChI Key | XKYFHUXZPRFUTH-UHFFFAOYSA-N |
Synonym | potassium manganate, unii-5pi213d3us, dipotassium dioxido dioxo manganese, acmc-1buxv, manganic acid h2mno4 , dipotassium salt, potassium manganate 10g, 2k.mno4, manganic acid h2mno4 , potassium salt 1:2 |
PubChem CID | 160931 |
IUPAC Name | dipotassium;dioxido(dioxo)manganese |
SMILES | [K+].[K+].[O-][Mn]([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11368308
|
Thermo Scientific Alfa Aesar
039506.09 |
10 g |
88.04 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11378308
|
Thermo Scientific Alfa Aesar
039506.18 |
50 g |
305.52 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It serves as an oxidizing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10294-64-1 | |
197.13 | |
XKYFHUXZPRFUTH-UHFFFAOYSA-N | |
160931 | |
[K+].[K+].[O-][Mn]([O-])(=O)=O |
K2MnO4 | |
MFCD00043089 | |
potassium manganate, unii-5pi213d3us, dipotassium dioxido dioxo manganese, acmc-1buxv, manganic acid h2mno4 , dipotassium salt, potassium manganate 10g, 2k.mno4, manganic acid h2mno4 , potassium salt 1:2 | |
dipotassium;dioxido(dioxo)manganese |
Specifications
190°C (decomposition) | |
Powder | |
MFCD00043089 | |
Soluble in water | |
[K+].[K+].[O-][Mn]([O-])(=O)=O | |
197.13 | |
197.13 |
10294-64-1 | |
K2MnO4 | |
potassium manganate, unii-5pi213d3us, dipotassium dioxido dioxo manganese, acmc-1buxv, manganic acid h2mno4 , dipotassium salt, potassium manganate 10g, 2k.mno4, manganic acid h2mno4 , potassium salt 1:2 | |
XKYFHUXZPRFUTH-UHFFFAOYSA-N | |
dipotassium;dioxido(dioxo)manganese | |
160931 | |
Potassium manganate |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: III
missing translation for 'einecsNumber' : 233-665-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Potassium manganate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
- MIXTURE LIST-Contains : Potassium manganate
RUO – Research Use Only