missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium metabisulfite, 97%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
Melting Point | 150.0°C |
Color | White to Yellow |
Physical Form | Fine Crystalline Powder |
Quantity | 25 g |
Assay Percent Range | 96% min. (Iodimetry) |
Linear Formula | K2S2O5 |
Merck Index | 15, 7766 |
Solubility Information | Solubility in water: 45g/L (20°C). Other solubilities: insoluble in ethanol |
Formula Weight | 222.33 |
Percent Purity | 97% |
Show More |
Safety and Handling
Product Identifier
- Potassium metabisulfite
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- EUH031-Contact with acids liberates toxic gas.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction