missing translation for 'onlineSavingsMsg'
Learn More

Potassium Metal In Liquid Paraffin, Pure, Fisher Chemical™

Product Code. 10121580
Click to view available options
Quantity:
10 g
100 g

Chemical Identifiers

CAS 7740-09-7
Molecular Formula K
MDL Number 133776

Specifications

Boiling Point 770°C
Melting Point 63°C
Color Gray
Physical Form Solid
Quantity 100 g
Formula Weight 39.1g/mol
Packaging Plastic container
Vapor Pressure 133mbar 171
compliance-icons compliance-icons
Product Identifier
  • Potassium, cubes, stored in mineral oil
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'