missing translation for 'onlineSavingsMsg'
Learn More
Potassium oxalate monohydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals Product Code.: 11356808

Potassium oxalate monohydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals

Product Code. 11356808
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11356808

Brand: Thermo Scientific Alfa Aesar 013452.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium oxalate monohydrate is used as a chelating agent; a purifying agent in pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, a rust remover for metal treatment, a bleaching agent in textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as a reducing agent in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
IUPAC Name dipotassium;oxalate;hydrate
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Melting Point >300°C
Color White
Density 2.13 g/mL
pH 6 to 8.5
Odor Odorless
Linear Formula K2C2O4·H2O
Quantity 100 g
Beilstein 3752576
Merck Index 14,7651
Solubility Information Soluble in water.
Formula Weight 184.23 (166.22 Anhydrous)
Percent Purity 98.8 to 101.0%
Grade ACS
Physical Form Powder or Granular Powder
Chemical Name or Material Potassium oxalate monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Potassium oxalate monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium oxalate monohydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.