missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium oxalate monohydrate, Honeywell™
Click to view available options
Quantity:
250 g
1 kg
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
Quantity | 250 g |
Linear Formula | (COOK)2 · H2O |
UN Number | NONH for all modes of transport |
Beilstein | 3752576 |
Formula Weight | 184.23g/mol |
Percent Purity | ≥99% (RT) |
Chemical Name or Material | Potassium oxalate monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'