Learn More
Potassium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals
25.53 € - 311.55 €
Chemical Identifiers
CAS | 7778-74-7 |
---|---|
Molecular Formula | ClKO4 |
Molecular Weight (g/mol) | 138.54 |
MDL Number | MFCD00011362 |
InChI Key | YLMGFJXSLBMXHK-UHFFFAOYSA-M |
Synonym | potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp |
PubChem CID | 516900 |
IUPAC Name | potassium;perchlorate |
SMILES | [K+].[O-][Cl](=O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10286320
|
Thermo Scientific Acros
221360250 |
25 g | Plastic bottle |
25.53 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10410881
|
Thermo Scientific Acros
221365000 |
500 g | Plastic bottle |
91.05 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10075363
|
Thermo Scientific Acros
221360010 |
1 kg | Plastic bottle |
156.78 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13438478
|
Thermo Scientific Acros
221360025 |
2.5 kg | Plastic bottle |
311.55 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7778-74-7 | |
138.54 | |
YLMGFJXSLBMXHK-UHFFFAOYSA-M | |
516900 | |
[K+].[O-][Cl](=O)(=O)=O |
ClKO4 | |
MFCD00011362 | |
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp | |
potassium;perchlorate |
Specifications
400.0°C | |
Colorless to Yellow | |
99+% | |
KClO4 | |
15,7774 | |
Solubility in water: 17g/L (20°C). Other solubilities: 0.75 g/100mL water (0°C), 21.8 g/100mL water (100°C), practically insoluble in alcohol | |
[K+].[O-][Cl](=O)(=O)=O | |
138.54 | |
138.55 | |
Analytical |
7778-74-7 | |
Liquid | |
ClKO4 | |
MFCD00011362 | |
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp | |
YLMGFJXSLBMXHK-UHFFFAOYSA-M | |
potassium;perchlorate | |
516900 | |
≥99% | |
Potassium perchlorate, p.a. |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause fire or explosion; strong oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/other combustible materials.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-912-9
missing translation for 'rtecsNumber' : SC9700000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.