missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Potassium permanganate, 99+%, for analysis
Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
Color | Green to Black |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 25 g |
Assay Percent Range | 99+% |
Linear Formula | KMnO4 |
Fieser | 01,942; 02,348; 04,412; 05,562; 08,416; 09,388; 10,330; 11,440; 12,413; 13,258; 14,267; 15,273; 16,283 |
Merck Index | 15, 7776 |
SMILES | [O-][Mn](=O)(=O)=O.[K+] |
IUPAC Name | potassium;permanganate |
Molecular Weight (g/mol) | 158.032 |
Show More |
Safety and Handling
Product Identifier
- Potassium permanganate
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'