missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium permanganate in aqueous solution, BAKER ANALYZED™, J.T. Baker™
Click to view available options
1 L
Specifications
Specifications
CAS | 7722-64-7 |
Quantity | 1 L |
Molecular Formula | KMnO4 |
MDL Number | MFCD00011364 |
InChI Key | VZJVWSHVAAUDKD-UHFFFAOYSA-N |
SMILES | [K+].[O-][Mn](=O)(=O)=O |
IUPAC Name | potassium manganesoylolate |
Molecular Weight (g/mol) | 158.03 |
Concentration or Composition (by Analyte or Components) | 0.02 mol/L (0.1 N) |
Packaging | Glass bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'