missing translation for 'onlineSavingsMsg'
Learn More

Potassium Permanganate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15568094
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

High quality chemicals for laboratory and specialized industrial use

    Suitable for Mercury Determination

Specifications

CAS 7722-64-7
Quantity 500 g
Molecular Formula KMnO4
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES [O-][Mn](=O)(=O)=O.[K+]
IUPAC Name potassium;permanganate
Molecular Weight (g/mol) 158.032
PubChem CID 516875
Formula Weight 158.03
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.70kg
Chemical Name or Material Potassium Permanganate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'