missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium permanganate solution, Volumetric, 0.1 M KMnO4 (0.5N), Honeywell Fluka™
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass bottle
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
Quantity | 2.5 L |
UN Number | UN3082 |
Formula Weight | 158.03g/mol |
Packaging | Glass bottle |
Chemical Name or Material | Potassium permanganate solution |
Safety and Handling
Product Identifier
- Potassium permanganate solution
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : potassium permanganate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'