missing translation for 'onlineSavingsMsg'
Learn More

Potassium peroxomonosulfate, extra pure, min. 4.5% active oxygen, Thermo Scientific Chemicals

Product Code. 10348490
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70693-62-8, 10058-23-8, 7778-80-5, 7646-93-7
Molecular Formula H3K5O18S4
Molecular Weight (g/mol) 614.78
MDL Number MFCD00040551
InChI Key HJKYXKSLRZKNSI-UHFFFAOYSA-I
Synonym Oxone, Potassium Monopersulfate

Specifications

Color White
Physical Form Granular Crystalline Powder
Quantity 100 g
Linear Formula KHSO4·K2SO4
Fieser 01,952; 12,413; 16,285
Solubility Information Solubility in water: 298g/L (20°C).
Formula Weight 614.78
Percent Purity ≥4.5% (active oxygen)
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Potassium peroxomonosulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium peroxomonosulfate compound, min. 4.5% active oxygen
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.