missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate ACS AR Crystal, Macron Fine Chemicals™

macron

147.00 € - 503.00 €

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium;dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15528754
View Documents
Avantor Macron
7100-12
500 g POLYSTORMOR
147.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15518754
View Documents
Avantor Macron
7100-06
2.5 kg Poly
503.00 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7778-77-0
136.08
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
potassium;dihydrogen phosphate
H2KO4P
MFCD00011401 MFCD00147253
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
CHEBI:63036
[K+].OP(O)([O-])=O

Specifications

7778-77-0
H2KO4P
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
[K+].OP(O)([O-])=O
136.08
CHEBI:63036
ACS
Crystalline
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium;dihydrogen phosphate
516951
136.09
Potassium Phosphate, Monobasic
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium Phosphate, Monobasic

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Potassium phosphate,monobasic

SDS
Documents

Documents

Product Certifications