missing translation for 'onlineSavingsMsg'
Learn More
Potassium phosphate, anhydrous, 97%, Thermo Scientific Chemicals Product Code.: 11378708

Potassium phosphate, anhydrous, 97%, Thermo Scientific Chemicals

Product Code. 11378708
250 g
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11378708

Brand: Thermo Scientific Alfa Aesar 041587.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium phosphate is used as a fertilizer and food additive. It serves as a buffering agent to regulate pH. It is also used in optical modulators and for non-linear optics such as second-harmonic generation (SHG). It is involved in the phase-transfer catalyzed preparation of acetylenes from the corresponding vicinal dibromo compounds through double dehydrobromination. It is also useful for the oxidative coupling of thiols to disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-53-2
Molecular Formula K3O4P
Molecular Weight (g/mol) 212.265
MDL Number MFCD00036295
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
Synonym potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321
PubChem CID 62657
IUPAC Name tripotassium;phosphate
SMILES [O-]P(=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Melting Point 1340°C
Physical Form Granules
Quantity 250 g
Assay Percent Range 97%
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 212.27
Sensitivity Hygroscopic
Density 2.564 g/mL
Chemical Name or Material Potassium phosphate, anhydrous

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW27K055 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog Number041587.A1,041587.30,041587.A7
Lot NumberM16K004 Certificate TypeCertificate of Analysis Date23/01/2024 Catalog Number041587.A1,041587.30,041587.A7
Lot NumberC13Z026 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number041587.A1,041587.30,041587.A7
Lot Number10187079 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041587.A1,041587.30,041587.A7
Lot NumberI10S013 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041587.A1,041587.30,041587.A7
compliance-icons compliance-icons
Product Identifier
  • Potassium phosphate, anhydrous
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Phosphoric acid, tripotassium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium phosphate, anhydrous, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.