missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, anhydrous, 97%, Thermo Scientific Chemicals

Product Code. 11388708
Click to view available options
Quantity:
250 g
1 kg
5 kg

Potassium phosphate is used as a fertilizer and food additive. It serves as a buffering agent to regulate pH. It is also used in optical modulators and for non-linear optics such as second-harmonic generation (SHG). It is involved in the phase-transfer catalyzed preparation of acetylenes from the corresponding vicinal dibromo compounds through double dehydrobromination. It is also useful for the oxidative coupling of thiols to disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-53-2
Molecular Formula K3O4P
Molecular Weight (g/mol) 212.265
MDL Number MFCD00036295
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
Synonym potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321
PubChem CID 62657
IUPAC Name tripotassium;phosphate
SMILES [O-]P(=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Melting Point 1340°C
Physical Form Granules
Quantity 5 kg
Merck Index 14,7660
Solubility Information Soluble in water; Insoluble in alcohol.
Formula Weight 212.27
Percent Purity 97%
Sensitivity Hygroscopic
Density 2.564
Chemical Name or Material Potassium phosphate, anhydrous

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'