missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic, ≥98%, ACS reagent, Honeywell Fluka™

Product Code. 15638020
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
4 x 2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-11-4
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
MDL Number MFCD00011383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Quantity 4 x 2.5 kg
Linear Formula K2HPO4
UN Number NONH for all modes of transport
Formula Weight 174.18g/mol
Percent Purity ≥98%
Chemical Name or Material Potassium phosphate dibasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'