missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate, dibasic, 98+%, ACS reagent, Thermo Scientific Chemicals
26.33 € - 711.54 €
Chemical Identifiers
CAS | 7758-11-4 |
---|---|
Molecular Formula | HK2O4P |
Molecular Weight (g/mol) | 174.18 |
MDL Number | MFCD00011383 |
InChI Key | ZPWVASYFFYYZEW-UHFFFAOYSA-L |
Synonym | dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate |
PubChem CID | 24450 |
ChEBI | CHEBI:32031 |
IUPAC Name | dipotassium;hydrogen phosphate |
SMILES | OP(=O)([O-])[O-].[K+].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10546982
|
Thermo Scientific Acros
424190050 |
5 g | Glass Bottle |
26.33 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10331211
|
Thermo Scientific Acros
424195000 |
500 g | Plastic bottle |
76.58 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10184333
|
Thermo Scientific Acros
424190025 |
2.5 kg | Plastic bottle |
236.18 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13438578
|
Thermo Scientific Acros
424190100 |
10 kg | Plastic Drum |
711.54 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-11-4 | |
174.18 | |
ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
24450 | |
dipotassium;hydrogen phosphate |
HK2O4P | |
MFCD00011383 | |
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
CHEBI:32031 | |
OP(=O)([O-])[O-].[K+].[K+] |
Specifications
340.0°C | |
White | |
Fine Crystalline Powder | |
HK2O4P | |
MFCD00011383 | |
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
dipotassium;hydrogen phosphate | |
24450 | |
174.18 | |
ACS Reagent | |
Potassium phosphate, dibasic |
7758-11-4 | |
8.5 to 9.6 (5% soln. at 25°C) | |
98+% | |
K2HPO4 | |
15,7779 | |
Solubility in water: 1600g/L (20°C). Other solubilities: slightly soluble in alcohol | |
OP(=O)([O-])[O-].[K+].[K+] | |
174.18 | |
CHEBI:32031 | |
≥98% | |
1.0% max. (105°C) |
Safety and Handling
HYGROSCOPIC
missing translation for 'einecsNumber' : 231-834-5
missing translation for 'tsca' : TSCA
RUO – Research Use Only