missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, ACROS Organics, Quantity: 5kg, Packaging: Plastic drum, CAS: 7758-11-4, White, Grade: Analysis, Melting Point: 340.0 deg.C, Molecular Formula: HK2O4P, Molecular Weight: 174.18, Percent Purity: 99+%, pH: 8.5 to 9.6 (5% soln. at 20 deg.C)

Chemical Identifiers

CAS 7758-11-4
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.18
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10509263
View Documents Promotion Details
Thermo Scientific Acros
215472500
250 g Plastic bottle N/A
N/A
10541081
View Documents Promotion Details
Thermo Scientific Acros
215470010
1 kg Plastic bottle N/A
N/A
11944291
View Documents Promotion Details
Thermo Scientific Acros
215470050
5 kg Plastic drum N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications

Chemical Identifiers

7758-11-4
174.18
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
CHEBI:32031
OP(=O)([O-])[O-].[K+].[K+]
HK2O4P
ZPWVASYFFYYZEW-UHFFFAOYSA-L
24450
dipotassium;hydrogen phosphate

Specifications

340.0°C
White
Fine Crystalline Powder
HK2O4P
15,7779
Solubility in water: 1600g/L (20°C). Other solubilities: slightly soluble in alcohol
OP(=O)([O-])[O-].[K+].[K+]
174.18
CHEBI:32031
≥99%
1% (130°C) max.
7758-11-4
8.5 to 9.6 (5% soln. at 20°C)
99+%
K2HPO4
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium;hydrogen phosphate
24450
174.18
Analytical
Potassium phosphate, dibasic
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-834-5

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only