missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate, dibasic trihydrate, 99+%, for analysis, Thermo Scientific Chemicals
9.40 € - 335.00 €
Chemical Identifiers
CAS | 16788-57-1 |
---|---|
Molecular Formula | H7K2O7P |
Molecular Weight (g/mol) | 228.22 |
MDL Number | MFCD00149926 |
InChI Key | XQGPKZUNMMFTAL-UHFFFAOYSA-L |
Synonym | dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate |
PubChem CID | 16217523 |
SMILES | O.O.O.[K+].[K+].OP([O-])([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10348580
|
Thermo Scientific Acros
205930250 |
25 g | Plastic bottle | ||||||
10226310
|
Thermo Scientific Acros
205935000 |
500 g | Plastic bottle |
106.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10732574
|
Thermo Scientific Acros
205930025 |
2.5 kg | Plastic bottle |
335.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
16788-57-1 | |
228.22 | |
XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
16217523 |
H7K2O7P | |
MFCD00149926 | |
dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
O.O.O.[K+].[K+].OP([O-])([O-])=O |
Specifications
16788-57-1 | |
8.5 to 9.6 (5% soln.) | |
99+% | |
K2HPO4·3H2O | |
10, 7541 | |
XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
dipotassium trihydrate hydrogen phosphate | |
16217523 | |
≥99.0% | |
Potassium phosphate, dibasic trihydrate, 99+% |
White | |
Adhering Crystals | |
H7K2O7P | |
MFCD00149926 | |
dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
O.O.O.[K+].[K+].OP([O-])([O-])=O | |
228.22 | |
228.23 | |
Analytical |
RUO – Research Use Only