missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Potassium phosphate, monobasic, 99+%, for biochemistry Product Code.: 12694957

Thermo Scientific Chemicals Potassium phosphate, monobasic, 99+%, for biochemistry

Product Code. 12694957
500 g
Click to view available options
Quantity:
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 12694957

Brand: Thermo Scientific Chemicals 212595000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253.0°C
Quantity 500 g
Assay Percent Range 99+%
Linear Formula KH2PO4
Merck Index 15,778
Solubility Information (10% in water) Clear colorless
IUPAC Name potassium dihydrogen phosphate
Formula Weight 136.09
Percent Purity ≥99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Potassium phosphate, monobasic
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467671 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number212590025,212595000
Lot NumberA0458976 Certificate TypeCertificate of Analysis Date20/12/2023 Catalog Number212590025,212595000
Lot NumberA0399560 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number212590025,212595000
Lot NumberA0411831 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number212590025,212595000
Lot NumberA0414373 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number212590025,212595000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Potassium phosphate, monobasic, 99+%, for biochemistry >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.