missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Monobasic, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
344.00 € - 562.00 €
Chemical Identifiers
CAS | 7778-77-0 |
---|---|
Molecular Formula | H2KO4P |
Molecular Weight (g/mol) | 136.08 |
MDL Number | MFCD00011401 MFCD00147253 |
InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphateShow More |
PubChem CID | 516951 |
ChEBI | CHEBI:63036 |
IUPAC Name | potassium dihydrogen phosphate |
SMILES | [K+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15108936
|
Avantor J.T.Baker
3246-01 |
500 g | Poly Bottle |
344.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15588094
|
Avantor J.T.Baker
3246-05 |
2.5 kg | Poly bottle |
562.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7778-77-0 | |
136.08 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
516951 | |
potassium dihydrogen phosphate |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphateShow More | |
CHEBI:63036 | |
[K+].OP(O)([O-])=O |
Specifications
7778-77-0 | |
H2KO4P | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
[K+].OP(O)([O-])=O | |
136.08 | |
CHEBI:63036 | |
Meets ACS reagent chemical requirements | |
Potassium Phosphate, Monobasic |
Crystals | |
MFCD00011401 MFCD00147253 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
potassium dihydrogen phosphate | |
516951 | |
136.09 | |
1L = 2.34kg |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Phosphate, Monobasic, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™