missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate monobasic, meets analytical spec. of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg
Beschreibung
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spezifikation
Spezifikation
CAS Max % | 100.0000% |
pH | 4.2 - 4.5 |
Quantity | 50 kg |
Assay Percent Range | 98 to 100.5% |
Linear Formula | KH2PO4 |
UN Number | NONH for all modes of transport |
Solubility Information | 4.5parts soluble in water, insoluble in alcohol |
IUPAC Name | potassium;dihydrogen phosphate |
Formula Weight | 136.09 |
Percent Purity | 98 to 100.5% (calc. to the dried substance) |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'