missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate monobasic, meets analytical spec. of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
CAS Max % | 100.0000% |
pH | 4.2 - 4.5 |
Quantity | 6 x 1 kg |
Assay Percent Range | 98 to 100.5% |
Linear Formula | KH2PO4 |
IUPAC Name | potassium;dihydrogen phosphate |
Formula Weight | 136.09 |
Grade | Reagent Ph. Eur. |
Loss on Drying | max. 1.0% |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'