missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate monobasic, meets analytical spec. of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15670720
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg

Meets analytical specification of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Physical Form Anhydrous
Quantity 1 kg
Linear Formula KH2PO4
UN Number NONH for all modes of transport
Solubility Information 4.5parts soluble in water, insoluble in alcohol
IUPAC Name potassium dihydrogen phosphate
Formula Weight 136.09g/mol
Percent Purity 98 to 100.5% (calc. to the dried substance)
Packaging Plastic Bottle
Density 2.338g/mL at 25°C
Chemical Name or Material Potassium Phosphate Monobasic, Meets analytical specification of Ph. Eur.NF
Show More Show Less
Product Identifier
  • Potassium phosphate monobasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'