missing translation for 'onlineSavingsMsg'
Learn More
Potassium phosphate, monobasic, for HPLC, Thermo Scientific Chemicals Product Code.: 16564204

Potassium phosphate, monobasic, for HPLC, Thermo Scientific Chemicals

Product Code. 16564204
50 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 16564204

Brand: Thermo Scientific Acros 447370500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253.0°C
Quantity 50 g
Linear Formula KH2PO4
Merck Index 15, 7780
Solubility Information Solubility aq. soln.: 222 g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 136.09
Grade HPLC
Chemical Name or Material Potassium phosphate, monobasic

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463588 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog Number447370010,447372500,447370500
Lot NumberA0450269 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number447370010,447372500,447370500
Lot NumberA0444102 Certificate TypeCertificate of Analysis Date14/07/2022 Catalog Number
Lot NumberA0441804 Certificate TypeCertificate of Analysis Date03/05/2022 Catalog Number447370010,447372500,447370500
Lot NumberA0436856 Certificate TypeCertificate of Analysis Date02/12/2021 Catalog Number447370010,447372500,447370500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium phosphate, monobasic, for HPLC, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.